Learn More
Medchemexpress LLC Poly ethylene glycol 500mg
SDP
Catalog No. 5000204966
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 10000) (PEGDA (MW 10000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 10000) can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-04966 Supplier Medchemexpress LLC Supplier No. HYW110542H500MG
Poly(ethylene glycol) diacrylate (MW 10000) (PEGDA (MW 10000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 10000) can be used in the formation of a cross-linked polymers[1]