Learn More
Medchemexpress LLC Poly ethylene glycol 5g
SDP
Catalog No. 5000205120
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 6000) (PEGDA (MW 6000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 6000) can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-05120 Supplier Medchemexpress LLC Supplier No. HYW110542G5G
Poly(ethylene glycol) diacrylate (MW 6000) (PEGDA (MW 6000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 6000) can be used in the formation of a cross-linked polymers[1]