Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) bis(2-ethylhexanoate)

Catalog No. 450596250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-059-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-059-6250ML Supplier Sigma Aldrich Supplier No. 450596250ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) bis(2-ethylhexanoate)

Specifications

CAS 9004-93-7
Melting Point -48°C
Density 1.02 g/mL (at 25°C)
Refractive Index n20/D 1.453 (literature)
Linear Formula CH3(CH2)3 CH(C2H5)CO(OCH2CH2)nO2CCH(C2H5)(CH2)3 CH3
Quantity 250 mL
MDL Number MFCD00241430
Synonym PEG bis(2-ethylhexanoate)
Molecular Weight (g/mol) 650
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.