Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)

Catalog No. 5423421KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-234-21KG 1 kg
54-234-2250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-234-21KG Supplier Sigma Aldrich Supplier No. 5423421KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pluronic(R) F108 is a tri block copolymer and a terminal primary dihydroxy-functional oligomer. It behaves as a nonionic surfactant. It is a surface active material whose properties are dependent on the hydrophilic (EO)/hydrophobic (PO) ratio. High EO content gives better o/w stabilizers.Pluronic(R) F108 was used as a surfactant in a medium to prevent the cells from sticking to the surface of a microfluidic device.{7}-{11}Pluronic may be potentially useful as a pathogen detector. It was used in the analysis of microbial community and genotyping single nucleotide polymorphisms (SNPs).{9} The triblock copolymer was used as a pore template for the fabrication of porous carbons and carbon/silica composite.{10}Pluronic is a registered trademark of BASF

Specifications

MDL Number MFCD00082049
Quantity 1 kg
Synonym Pluronic™ F-108; PEG-PPG-PEG
Molecular Weight (g/mol) 14600
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.