Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene Glycol)-block-poly(propylene Glycol)-block-poly(ethylene Glycol) Diacrylate Average Mn~14 600

Catalog No. 9146651G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-466-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-466-51G Supplier Sigma Aldrich Supplier No. 9146651G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poloxamers are nonionic, triblock copolymers containg a central hydrophobic chain of poly(propylene glycol) (PPG) and two terminal hydrophilic chains of poly(ethylene glycol) (PEG). Due to their amphiphilic structure, poloxamer solutions feature temperature dependent self-assembling and thermo-gelling behavior. Concentrated aqueous solutions of poloxamers are liquid at low temperature and form a gel at higher temperature, in a reversible process. This characteristic has allowed for these materials to be used as drug carriers for most routes of administration including oral, topical, intranasal, ocular, and parenteral. Acrylate-modified derivatives are most commonly used as thermosensitive hydrogels for drug delivery and 3D bioprinting applications. After casting or printing, the material can be crosslinked to preserve the hydrogel structure.

Specifications

Linear Formula C3H3O[C2H4O]x[C3H6O]y[C2H4O]zC3H3O2
Quantity 1 g
Synonym F-108 diacrylate; PEG-PPG-PEG; Pluronic diacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.