Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) diacrylate

Catalog No. 475629500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-562-9500ML 500 mL
47-562-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-562-9500ML Supplier Sigma Aldrich Supplier No. 475629500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) diacrylate (PEGDA) is a polyethylene glycol (PEG) based material that can be used for a variety of tissue engineering and drug delivery based applications. It is majorly used as a prepolymer solution that can be used in the formation of a cross-linked polymeric system.PEGDA can be used in the formation of a UV-cured membrane for potential usage in the separation of carbon dioxide (CO2) based gases. It may also be used in the development of novel injectable biodegradable polymers for a variety of biomedical applications.

Specifications

CAS 26570-48-9
Density 1.11 g/mL (at 25°C)
Refractive Index n20/D 1.463
MDL Number MFCD00081876
Quantity 500 mL
Synonym PEG diacrylate
Molecular Weight (g/mol) 250
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.