Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) diamine

Catalog No. 7524605G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-246-05G 5 g
75-246-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-246-05G Supplier Sigma Aldrich Supplier No. 7524605G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) diamine

Specifications

CAS 24991-53-5
Melting Point 56°C to 60°C
Linear Formula H2N(CH2CH2O)nCH2CH2NH2
Quantity 5 g
MDL Number MFCD00132004
Synonym Poly(ethylene glycol) bis(amine); O,O-Bis(2-aminoethyl)polyethylene glycol; Diaminopolyethylene glycol; PEG-diamine; Polyoxyethylene bis(amine)
Molecular Weight (g/mol) 10000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.