Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) dibenzoate

Catalog No. 458074500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-807-4500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-807-4500ML Supplier Sigma Aldrich Supplier No. 458074500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Plasticizer

Specifications

CAS 9004-86-8
Density 1.16 g/mL (at 25°C)
Boiling Point 217°C (1 mmHg)
Refractive Index n20/D 1.528 (literature)
Linear Formula C7H5O(C2H4O)nC7H5O2
MDL Number MFCD00278885
Quantity 500 mL
Synonym Polyethylene glycol dibenzoate; Polyoxyethylene dibenzoate
Molecular Weight (g/mol) 410
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.