Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol) for GPC 108,000, MilliporeSigma™ Supelco™

Catalog No. 111013722 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3722 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3722 Supplier MilliporeSigma Supelco Supplier No. 81201500MG
Add to Cart
Add to Cart

Poly(ethylene glycol) is the most widely used non-ionic, hydrophilic polymer, which can find wide applications in biomedical industry such as in drug delivery systems

TRUSTED_SUSTAINABILITY

Specifications

CAS 25322-68-3
Flash Point 229°C
Physical Form Neat
Linear Formula H(OCH2 CH2)nOH
MDL Number MFCD00081839
Synonym PEG
Type Analytical Standard
Quantity 500 mg
Grade GPC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.