Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) methacrylate

Catalog No. 409537500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-953-7500ML 500 mL
40-953-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-953-7500ML Supplier Sigma Aldrich Supplier No. 409537500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polyethylene glycol (PEG) is a suitable material for biological applications since it does not trigger an immune response. It can by synthesized by anionic ring opening polymerization of ethylene oxide with a range of end groups (such as methacrylate: PEGMA) attached. PEG is a hydrophilic polymer that can form hydrogels when cross linked into networks. Acrylate and methacrylate chain ends undergo chain polymerization to from PEGMA hydrogels.

Specifications

CAS 25736-86-1
Density 1.105 g/mL (at 25°C)
Refractive Index n20/D 1.464
Linear Formula H2C=C(CH3)CO(OCH2CH2)nOH
Quantity 500 mL
MDL Number MFCD00081879
Synonym Ethoxylated 2-hydroxyethyl methacrylate; PEGMA; Poly(ethylene oxide) monomethacrylate
Molecular Weight (g/mol) 360
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.