Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) methyl ether

Catalog No. 202509500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-250-9500G 500 g
20-250-95G 5 g
20-250-9250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-250-9500G Supplier Sigma Aldrich Supplier No. 202509500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

It is a Polyethylene glycol (PEG) macromer with a reactive chain end consisting of methyl ether. Etherification of the PEG chain ends can be undertaken in basic conditions by reacting it with alkyl halides. mPEG can undergo cross linking to form hydrogels; polymerization can be initiated by redox reaction or free radical initiator.

Specifications

CAS 9004-74-4
Linear Formula CH3(OCH2CH2)nOH
Quantity 500 g
MDL Number MFCD00084416
Synonym Methoxy poly(ethylene glycol); Polyethylene glycol monomethyl ether; mPEG
Molecular Weight (g/mol) 2000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.