Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) methyl ether methacrylate

Catalog No. 447943100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-794-3100ML 100 mL
44-794-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-794-3100ML Supplier Sigma Aldrich Supplier No. 447943100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) methyl ether methacrylate

Specifications

CAS 26915-72-0
Melting Point -1°C to +2°C
Density 1.08 g/mL (at 25°C)
Refractive Index n20/D 1.496
Linear Formula H2C=CCH3CO2(CH2CH2O)nCH3
Quantity 100 mL
MDL Number MFCD00241432
Synonym Methoxy PEG methacrylate; Methoxy poly(ethylene glycol) monomethacrylate; Poly(ethylene glycol) monomethyl ether monomethacrylate
Molecular Weight (g/mol) 500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.