Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol methyl ether), average M.W. 750

Catalog No. AC192320010
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192320010 1 kg Glass bottle
AC192325000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC192320010 Supplier Thermo Scientific Chemicals Supplier No. 192320010
Only null left
Add to Cart

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-74-4
Molecular Formula (C2H4O)nCH4O
Molecular Weight (g/mol) 76.10
MDL Number MFCD00084416
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
SMILES COCCO

Specifications

Color White
pH 4 to 7 (5% aq. soln.)
Physical Form Fused Mass
Avg. Mol. Wt. or Mol. Wt. Range 715 to 785
Linear Formula CH3(OCH2CH)nOH
Density 1.09g/mL
Quantity 1 kg
IUPAC Name 2-methoxyethanol
Hydroxyl Value 71.5 to 78.5mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.1% max.
Chemical Name or Material Poly(ethylene glycol methyleether), Average M.W. 750
Show More Show Less
RTECSNumber PE8263000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.