Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether N'-(3-maleimidopropionyl)aminoethane

Catalog No. 757853100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-785-3100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-785-3100MG Supplier Sigma Aldrich Supplier No. 757853100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PEG average Mn 3,400; n∼77Applications have been reported for: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization.

Specifications

CAS 851040-94-3
Linear Formula C7H7N2O3(C2H4O)nC9H12NO4
Quantity 100 mg
Synonym Maleimide polyoxyethylene NHS-protected valeric acid; Maleimide-PEG-NHS protected acid; Maleimide-PEG-acid; maleimide-PEG-valeric acid
Molecular Weight (g/mol) 4000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.