Learn More
Medchemexpress LLC Poly(ethylene glycol) phenyl ether acrylate | 56641-05-5 | MFCD00197904 | 98.0% | 324 (average) | CH2CHCO2(CH2CH2O)nC6H5 | 10mL
SDP
Catalog No. 5000270128
Encompass_Preferred
Poly(ethylene glycol) phenyl ether acrylate (MW 324) is a difunctional cross-linking agent[1]
Catalog No. 50-002-70128 Supplier Medchemexpress LLC Supplier No. HYW76359710ML
Poly(ethylene glycol) phenyl ether acrylate (MW 324) is a difunctional cross-linking agent[1]