Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) phenyl ether acrylate

Catalog No. 407348100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-734-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-734-8100ML Supplier Sigma Aldrich Supplier No. 407348100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly (ethylene glycol) phenyl ether acrylate is hetero-bi functionalized. MEHQ inhibitor prevents auto polymerization.

Specifications

CAS 56641-05-5
Density 1.127 g/mL (at 25°C)
Boiling Point 134°C (1 mmHg)
Refractive Index n20/D 1.502
Linear Formula CH2CHCO2(CH2CH2O)nC6H5
MDL Number MFCD00197904
Quantity 100 mL
Synonym Ethoxylated phenyl acrylate; Phenol ethoxylate acrylate
Molecular Weight (g/mol) 324
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.