Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene oxide)

Catalog No. 1894645G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-946-45G 5 g
18-946-4250G 250 g
18-946-4500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-946-45G Supplier Sigma Aldrich Supplier No. 1894645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly (ethylene oxide) (PEO) is a crystalline thermoplastic polymer. It is usually available over a wide range of molecular weights; the higher molecular weight entities are called poly (ethylene oxide), poly-oxyethylene or polyoxirane.PEO may be used to afford PEO-salt complexes which are act as polymeric electrolytes to be used for alkali metal rechargeable batteries. PEO-polyester micelles have been reportedly used for drug delivery. PEO hydrogels have been reported to be used for culture media, replacing agar. Cross-linked PEO may be used for drug delivery and blood filtration. Compounds form association complexes with high molecular weight PEO; for example:bromine combines with PEO in controlled release, carboxymethyl dextran and carechol tannis form complexes with PEO to act as adhesives, copolymer of methyl vinyl ether and maleic anhydride form complex with PEO for its use in microencapsulation of non aqueous inks.Industrially, PEO may be used to modify bituminous emulsion in paving compositions and as a dispersant for calamine lotion.

Specifications

CAS 25322-68-3
Quantity 5 g
Linear Formula (-CH2CH2O-)n
MDL Number MFCD00081839
Synonym PEO
Molecular Weight (g/mol) 4000000
Density 1.21 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.