Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene oxide)

Catalog No. 3727815G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-278-15G 5 g
37-278-1500G 500 g
37-278-1250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 37-278-15G Supplier Sigma Aldrich Supplier No. 3727815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene oxide) (PEO) is a crystalline, thermoplastic, water-soluble polymer that is commercially available.PEO (Mv ∼ 1,000,000, powder) can be used in the fabrication of hollow TiO2 fibers, which can be used for a variety of applications such as drug delivery, catalysts, sensors, and electric devices. It can also be used as an electrolyte for the formation of dye-sensitized solar cells (DSSCs).

Specifications

CAS 25322-68-3
Quantity 5 g
Linear Formula (-CH2CH2O-)n
MDL Number MFCD00081839
Synonym PEO
Molecular Weight (g/mol) 1000000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.