Learn More
Description
- Heat-sealable or compatible with evidence security tape
- Secures all types of evidence
- Tamper-evident, Moisture-resistant, and durable
- Crafted from 4 mm polyethylene
- Available in multiple sizes, they can be Heat-sealed or secured with evidence tape for maximum integrity.
Specifications
Specifications
| Dimensions (L x W) | 20 x 30 in. |
| Product Type | Evidence Bag |
| Color | Transparent |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
