Learn More
McMaster-Carr Polyurethane Foam Clean Room Swabs, 1/8" Diameter x 13/32" Long Tip, 2-3/4" Overall Length, 500/pk
Catalog No. NC1441468
Encompass
Not only are these swabs adhesive and lint free, they also have the best solvent-holding ability of any swab.
Catalog No. NC1441468 Supplier McMaster-Carr Supplier No. 6111T2
Not only are these swabs adhesive and lint free, they also have the best solvent-holding ability of any swab.