Learn More
MSC Showa Cut-Resistant Gloves, HPPE, Polyurethane Coated, Size Small, 6/Pk.
Catalog No. 5000687178
Encompass
The 546 series gloves are designed to provide superior cut, puncture, and abrasion resistance for diverse industrial tasks. Featuring an ergonomic, seamless-knit construction with a high-performance polyethylene (HPPE) reinforced liner, these gloves ensure both protection and tactile precision. The palm and fingertips are coated with foamed polyurethane to deliver an excellent grip while maintaining breathability and flexibility for long-term comfort.
  • Engineered with a 13-gauge seamless-knit HPPE liner for cut protection
  • Foamed polyurethane coating on palm and fingertips for enhanced grip
  • Ergonomic design ensures high flexibility and dexterity for precision handling
  • Breathable construction supports extended wear
  • Machine washable for repeated use
  • ANSI/ISEA cut resistance level A3
  • Suitable for mechanical engineering, automotive repair, and handling sharp-edged objects
Catalog No. 50-006-87178 Supplier MSC Supplier No. 07915184
The 546 series gloves are designed to provide superior cut, puncture, and abrasion resistance for diverse industrial tasks. Featuring an ergonomic, seamless-knit construction with a high-performance polyethylene (HPPE) reinforced liner, these gloves ensure both protection and tactile precision. The palm and fingertips are coated with foamed polyurethane to deliver an excellent grip while maintaining breathability and flexibility for long-term comfort.
  • Engineered with a 13-gauge seamless-knit HPPE liner for cut protection
  • Foamed polyurethane coating on palm and fingertips for enhanced grip
  • Ergonomic design ensures high flexibility and dexterity for precision handling
  • Breathable construction supports extended wear
  • Machine washable for repeated use
  • ANSI/ISEA cut resistance level A3
  • Suitable for mechanical engineering, automotive repair, and handling sharp-edged objects