Learn More
MSC Chapin Sprayer, 1 gal, Polyethylene Tank, Coiled Hose, 1 ea
Catalog No. 502867192
Encompass
This 1 gal. garden hand sprayer features a durable polyethylene tank and a coiled hose, designed for efficient application of foaming solutions and general-purpose cleaning tasks.
  • Handheld sprayer type
  • Polyethylene tank material
  • Volume capacity of 1 gal.
  • Foam spray pattern
  • Not chemical safe
  • Anti-clog filter
  • Pressure relief valve
  • Ergonomic handle and coiled hose
  • Nozzle sprays a consistent foam stream
  • Wand length of 12 in
  • Coiled hose length of 10 ft (120 in)
  • Opening diameter of 3 in
Catalog No. 50-286-7192 Supplier MSC Supplier No. 02399418
This 1 gal. garden hand sprayer features a durable polyethylene tank and a coiled hose, designed for efficient application of foaming solutions and general-purpose cleaning tasks.
  • Handheld sprayer type
  • Polyethylene tank material
  • Volume capacity of 1 gal.
  • Foam spray pattern
  • Not chemical safe
  • Anti-clog filter
  • Pressure relief valve
  • Ergonomic handle and coiled hose
  • Nozzle sprays a consistent foam stream
  • Wand length of 12 in
  • Coiled hose length of 10 ft (120 in)
  • Opening diameter of 3 in