Learn More
Molecular Dimensions 20% w/v γ-PGA (Na+ form, LM), 100mL
SDP
Supplier:  Molecular Dimensions MD2100108
Encompass
20% w/v γ-PGA (Na+ form, LM) cosmetic grade (γ-Polyglutamic acid LM) average MW ~200-400kDa. Made with ultra-purified water and filter sterilized.
Catalog No. NC0022749