Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(hexamethylene diisocyanate)

Catalog No. 418005100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-800-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-800-5100ML Supplier Sigma Aldrich Supplier No. 418005100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(hexamethylene diisocyanate) (PolyHDI) is an aliphatic compound belonging to the class of isocyanates. It is generally prepared by reacting hexamethylene diamine with phosgene.PolyHDI can be used as a compatibilizer to increase the complex viscosity of poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV)/poly(lactic acid) biopolymer based blends. It can be used as a hardener that can be mixed with polyol resins to form polyurethane coatings with a wide range of applications like self-cleaning.

Specifications

CAS 28182-81-2
Density 1.12 g/mL (at 25°C)
Refractive Index n20/D 1.502
MDL Number MFCD00197906
Quantity 100 mL
Synonym 1,6-Diisocyanatohexane homopolymer; 1,6-Diisocyanatohexane polymer; HMDI homopolymer
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.