Learn More
Medchemexpress LLC Poly(hexamethylenebiguanide) hydrochloride | 32289-58-0 | MFCD00217750 | 98.0% | 213.33 g/mol | (C8H17N5)n.xHCl | 25 G
SDP
Catalog No. 5000303830
Encompass_Preferred
Poly(hexamethylenebiguanide) hydrochloride (PHMB) is a polymeric antiseptic and antiviral agent for research use. It is active against Gram-positive and Gram-negative bacteria and acts by adsorbing to surfaces and disrupting microbial cell membranes and metabolism. Common applications include medical textiles, household textiles, and cosmetics.
  • Broad-spectrum antimicrobial and antiviral activity.
  • Solid, white to off-white material for convenient handling.
  • High supplied purity: 98.0%.
  • Polymeric formula: (C8H17N5)n.xHCl for repeat-unit characterization.
  • Available in small research sizes for laboratory use.
Catalog No. 50-003-03830 Supplier Medchemexpress LLC Supplier No. 25GHYW017766
Poly(hexamethylenebiguanide) hydrochloride (PHMB) is a polymeric antiseptic and antiviral agent for research use. It is active against Gram-positive and Gram-negative bacteria and acts by adsorbing to surfaces and disrupting microbial cell membranes and metabolism. Common applications include medical textiles, household textiles, and cosmetics.
  • Broad-spectrum antimicrobial and antiviral activity.
  • Solid, white to off-white material for convenient handling.
  • High supplied purity: 98.0%.
  • Polymeric formula: (C8H17N5)n.xHCl for repeat-unit characterization.
  • Available in small research sizes for laboratory use.