Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(hexyl methacrylate) solution

Catalog No. 18212525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-212-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-212-525G Supplier Sigma Aldrich Supplier No. 18212525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(hexadecyl methacrylate) belongs to a class of long chain of poly (alkyl) methacrylate. It can be dissolved in benzene and heptane.Poly(hexyl methacrylate)(PHMA) may be used to prepare amphiphilic block copolymers with -poly(ethylene oxide) (PEO). These block copolymers may be used as structure directing agents for the preparation organic-inorganic hybrid materials from organic, volatile solvents.Unit weight does not include weight of solvent.

Specifications

CAS 25087-17-6
Density 1.007 g/mL (at 25°C)
Linear Formula (C10H18O2)n
Refractive Index n20/D 1.481
Quantity 25 g
MDL Number MFCD00084433
Synonym Hexyl methacrylate homopolymer; Poly(1-hexyl methacrylate)
Molecular Weight (g/mol) 400000
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.