Learn More
Bioworld Anti-Poly-His Tag Antibody (Rabbit Polyclonal IgG), 200 μg
Supplier:  Bioworld 203120991
Encompass_Preferred
Anti-Poly-His tag antibody (Rabbit Polyclonal IgG)Cat#20312099 DetailsHost: RabbitIsotype: IgGClass: PolyclonalImmunogen: Synthetic peptide corresponding to HHHHHH conjugated to KLH as the immunogenConjugate: UnconjugatedForm InformationForm: LiquidConcentration: 2 mg/mLPurification: Protein A ChromatrographyStorage Buffer: PBS with 0.1% BSAPreservative: Upon Request (0.02% Sodium Azide)Storage:Store at +4°C short term (1-2 weeks). Aliquot and store at -20°C or -80°C. Avoid repeated freeze / thaw cycles.Usage RecommendationsWestern Blot (WB): 1:3000-1:5000ELISA: 1:150-1:400Immunoprecipitation (IP): 1:350 - 1:500
Catalog No. 50-198-021