Learn More
Cole-Parmer Advantec 43303020 Polypropylene Filter Holder for 47 mm Membranes
Supplier:  Cole-Parmer 0662322
Encompass
Use these polypropylene holders for ultracleaning and sterilizing liquids under pressure, or aseptic sampling of liquids and gases. Its maximum operating temperature for liquids is 176>F or 80>C.
Catalog No. NC9974894