Learn More
MSC Buyers Products SaltDogg Poly Hopper Spreader, 4 cubic yard, Auger
Catalog No. 502839355
Encompass
This hopper spreader is designed for medium and heavy-duty commercial use by contractors and property managers.
  • 4 cubic yard poly hopper resists rust and dents.
  • Handles bulk salt or 50/50 sand/salt mix.
  • Available in 12V DC-powered electric or hydraulic drive.
  • Independent auger and spinner motors.
  • 5-1/2 inch steel auger with stainless steel trough.
  • 400 lb. vibrator prevents material clumping.
  • 18 inch polymer spinner broadcasts material up to 30 ft.
  • In-cab control for independent auger/spinner speeds and vibrator (electric version).
  • Removable spinner assembly for cleaning.
  • Includes top screens, inverted V, tie downs, and auger grease kit.
  • 2-year warranty.
  • Made in USA.
Catalog No. 50-283-9355 Supplier MSC Supplier No. 77239853
Add to Cart
Add to Cart
Request a Quote
This hopper spreader is designed for medium and heavy-duty commercial use by contractors and property managers.
  • 4 cubic yard poly hopper resists rust and dents.
  • Handles bulk salt or 50/50 sand/salt mix.
  • Available in 12V DC-powered electric or hydraulic drive.
  • Independent auger and spinner motors.
  • 5-1/2 inch steel auger with stainless steel trough.
  • 400 lb. vibrator prevents material clumping.
  • 18 inch polymer spinner broadcasts material up to 30 ft.
  • In-cab control for independent auger/spinner speeds and vibrator (electric version).
  • Removable spinner assembly for cleaning.
  • Includes top screens, inverted V, tie downs, and auger grease kit.
  • 2-year warranty.
  • Made in USA.