Learn More
Invivogen Poly(I:C) HMW, Long Synthetic Analog of dsRNA, 10 mg
SDP
Catalog No. NC9180242
Encompass
Poly(I: C), Synthetic analog of double stranded RNA, 25ug/mL, Polyinosine-polycytidylic acid (poly(I: C)) is a synthetic analog of double-stranded RNA (dsRNA), a molecular pattern associated with viral infection, TLR3 Ligand, 10mg
Catalog No. NC9180242 Supplier Invivogen Supplier No. TLRLPIC
Poly(I: C), Synthetic analog of double stranded RNA, 25ug/mL, Polyinosine-polycytidylic acid (poly(I: C)) is a synthetic analog of double-stranded RNA (dsRNA), a molecular pattern associated with viral infection, TLR3 Ligand, 10mg