Learn More
Invivogen POLY(I:C) 50MG
SDP
Catalog No. NC9845570
Encompass
Long synthetic analog of dsRNA Polyinosinic-polycytidylic acid (poly(I:C)) is a synthetic analog of double-stranded RNA (dsRNA), a molecular pattern associated with viral infection. Poly(I:C) is recognized by TLR3 inducing the activation of NF-kB and the production of cytokines. Working concentration: 30 ng- 10 ug/ml Endotoxin level: <0.001 EU/ug Solubility: 5 mg/ml in physiological water (NaCl 0.9%) heated for 10 minutes at 65 - 70 Deg.C
Catalog No. NC9845570 Supplier Invivogen Supplier No. TLRLPIC5
Long synthetic analog of dsRNA Polyinosinic-polycytidylic acid (poly(I:C)) is a synthetic analog of double-stranded RNA (dsRNA), a molecular pattern associated with viral infection. Poly(I:C) is recognized by TLR3 inducing the activation of NF-kB and the production of cytokines. Working concentration: 30 ng- 10 ug/ml Endotoxin level: <0.001 EU/ug Solubility: 5 mg/ml in physiological water (NaCl 0.9%) heated for 10 minutes at 65 - 70 Deg.C