Learn More
Enzo Life Sciences Polyinosinic-polycytidylic acid. potassium salt (TLRGRADE™) (synthetic) (2mg). CAS: 31852-29-6
SDP
Catalog No. 502009664
Encompass_Preferred
Specific activator for TLR3 (Toll-like receptor 3) and MDA5/Helicard. Alternative name: poly(I:C) . K. Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.
Catalog No. 50-200-9664 Supplier Enzo Life Sciences Supplier No. ALX746021M002
Specific activator for TLR3 (Toll-like receptor 3) and MDA5/Helicard. Alternative name: poly(I:C) . K. Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.