Learn More
Medchemexpress LLC Poly (I:C): kanamycin (1:1) sodium | 5 MG
SDP
Catalog No. 5000222355
Encompass_Preferred
Poly (I:C):Kanamycin (1:1) (sodium) is a 1:1 isometric complex of the synthetic double-stranded RNA analog polyinosinic-polycytidylic acid and kanamycin (sodium). The complex functions as a TLR3 and RIG-I/MDA5 agonist to activate innate and adaptive immune responses and is used as an immunostimulant and vaccine adjuvant in research. Recommended storage: -20°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Acts as TLR3 and RIG-I/MDA5 agonist
  • Used as immunostimulant and vaccine adjuvant in research
  • Includes kanamycin component with antibacterial activity
  • Provided as a sodium salt complex for stability
  • Store sealed at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month)
Catalog No. 50-002-22355 Supplier Medchemexpress LLC Supplier No. HY135748A5MG
Poly (I:C):Kanamycin (1:1) (sodium) is a 1:1 isometric complex of the synthetic double-stranded RNA analog polyinosinic-polycytidylic acid and kanamycin (sodium). The complex functions as a TLR3 and RIG-I/MDA5 agonist to activate innate and adaptive immune responses and is used as an immunostimulant and vaccine adjuvant in research. Recommended storage: -20°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Acts as TLR3 and RIG-I/MDA5 agonist
  • Used as immunostimulant and vaccine adjuvant in research
  • Includes kanamycin component with antibacterial activity
  • Provided as a sodium salt complex for stability
  • Store sealed at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month)