Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Poly(I:C) Sodium Salt, Lyophilized, >98% Purity, 25 mg
SDP

Catalog No. 502050568
Encompass

Poly(I:C) Sodium Salt is a synthetic analog to double-stranded RNA (dsRNA) and a potent activator of Toll-like receptor 3 (TLR3). It plays a significant role in the innate immune response by mimicking viral infection to trigger the activation of transcription factors NF-κB and IRF3. This reagent is intended for research use.

  • Acts as a potent TLR3 agonist
  • Supplied as a lyophilized powder
  • Purity of >98%
  • Soluble in water at 10 mg/mL with warming
  • Compatible with use in studies investigating vaccine immunogenicity

Catalog No. 50-205-0568 Supplier Cell Signaling Technology Supplier No. 61401S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly(I:C) Sodium Salt is a synthetic analog to double-stranded RNA (dsRNA) and a potent activator of Toll-like receptor 3 (TLR3). It plays a significant role in the innate immune response by mimicking viral infection to trigger the activation of transcription factors NF-κB and IRF3. This reagent is intended for research use.

  • Acts as a potent TLR3 agonist
  • Supplied as a lyophilized powder
  • Purity of >98%
  • Soluble in water at 10 mg/mL with warming
  • Compatible with use in studies investigating vaccine immunogenicity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.