Learn More
Alamanda Polymers POLY-L-GLUTAMIC ACID SODI100MG
SDP
Catalog No. NC2942772
Encompass
poly-L-glutamic acid sodium salt, MW=15,000 Da), 100mg
Catalog No. NC2942772 Supplier Alamanda Polymers Supplier No. PLE100100MG
poly-L-glutamic acid sodium salt, MW=15,000 Da), 100mg