Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(L-lactide-co-glycolide)

Catalog No. 9010215G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-102-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-102-15G Supplier Sigma Aldrich Supplier No. 9010215G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(L-lactide-co-glycolide) (PLGA) is a biodegradable and biocompatible copolymer synthesized from the ring opening polymerization of lactide and glycolide. Due to the ester linkages found in the polymer backbone, these polymers are able to degrade via hydrolysis. Depending on the ratio of monomers used and polymer block structure, different forms of PLGA can be obtained. These differing forms yield materials with different degrees of crystallinity, thermal transitions, solubility, and rates of degradation.Poly(L-lactide-co-glycolide) (PLGA) is a biodegradable and biocompatible copolymer for use in biomedical research devices, such as grafts, sutures, implants, prosthetic devices, surgical sealant films, scaffolds for tissue engineering applications, micro and nanoparticles. PLGA is also utilized in drug delivery research, such as in the synthesis and application of polymer-drug conjugates.

Specifications

Linear Formula (C4H4O4)m(C6H8O4)n
Quantity 5 g
Synonym PLGA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.