Learn More
Newcomer Supply Poly-L-Lysine Adhesive Stock, 100 ml
SDP
Catalog No. NC9663893
Encompass
Newcomer Supply Poly-L-Lysine Adhesive Stock when used as a diluted working solution, provides a strong adhesive coating to microscopic slides.  This applied coating is very effective to further enhance bonding of tissue sections to slides for use in histological, microwave and immunohistochemistry (IHC) staining procedures and will leave very minimal or no background staining. One liter of Poly-L-Lysine Working Solution will coat approximately 900 slides. Exceeding 900 slides per liter of working solution may affect product performance. Shelf Life is 1 year from date of manufacture. Store at 2-8°C.
Catalog No. NC9663893 Supplier Newcomer Supply Supplier No. 1339A
Newcomer Supply Poly-L-Lysine Adhesive Stock when used as a diluted working solution, provides a strong adhesive coating to microscopic slides.  This applied coating is very effective to further enhance bonding of tissue sections to slides for use in histological, microwave and immunohistochemistry (IHC) staining procedures and will leave very minimal or no background staining. One liter of Poly-L-Lysine Working Solution will coat approximately 900 slides. Exceeding 900 slides per liter of working solution may affect product performance. Shelf Life is 1 year from date of manufacture. Store at 2-8°C.