Learn More
Sciencell Research Laboratories POLY-L-LYSINE 1MG/ML 1MLPOLY-
Catalog No. NC9664286
Encompass
Poly-L-Lysine is a synthetic compound that is a highly positively charged amino acid chain. It is commonly used as a coating agent to promote cell adhesion and improve the survival of primary cells in culture.
Catalog No. NC9664286 Supplier Sciencell Research Laboratories Supplier No. 0403
Poly-L-Lysine is a synthetic compound that is a highly positively charged amino acid chain. It is commonly used as a coating agent to promote cell adhesion and improve the survival of primary cells in culture.