Learn More
Neuvitro Poly-L-lysine coated sterile German glass coverslips for cell culture, 22mm dia #1 thick 90/pk, ready to use in cell culture.
Catalog No. NC0585187
Encompass_Preferred
CE certified German glass coverslips with Poly-L-lysine coating on both sides for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, coated with synthetic Poly-L-lysine on both sides and sterile. 22mm diameter round, #1 thickness, 90 pieces/pack. Ready to use directly in cell culture. Shelf life 0.5 year when stored in chest freezer. User manual included.
Catalog No. NC0585187 Supplier Neuvitro Supplier No. GG22PLL
CE certified German glass coverslips with Poly-L-lysine coating on both sides for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, coated with synthetic Poly-L-lysine on both sides and sterile. 22mm diameter round, #1 thickness, 90 pieces/pack. Ready to use directly in cell culture. Shelf life 0.5 year when stored in chest freezer. User manual included.