Learn More
Polysciences, Inc. Poly(l-lysine hydrobromide), 25988-63-0, MFCD00237230, 50 mg
SDP
Supplier:  Polysciences, Inc. 1861950
Encompass
Molecular Formula: C18 H36 N6 O3 X2 . 3 Br H, Molecular Weight: 627.257. Poly-L-lysine (PLL) hydrobromide is synthetic cationic, positively charged, amino acid polymer. It is used for promotion of cell adhesion to solid surfaces, e.g., Polysciences product # 22247, PLL coated microscope slides provide superior cell and tissue adhesion allowing for easier manipulation of samples. Appearance: powder, Handling: Gloves & fume hood, Storage: Store at room temperature.
Catalog No. NC9505830