Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Poly-L-lysine hydrobromide | 25988-63-0 | 70000-150000 | 50 ML
SDP

Catalog No. 5000436248
Encompass_Preferred

Poly-L-lysine solution (0.01%, sterile-filtered, MW 70000-150000) is a biochemical reagent intended for research use only, and not for sale to patients. This solution is also referred to as PLL solution 0.01%, sterile-filtered, hydrobromide, MW 70000-150000, and is suitable for cell culture applications.

  • Molecular weight ranges from 70000 to 150000
  • Chemical formula is (C6H14N2O2)n.nBrH
  • CAS number is 25988-63-0
  • Appears as a liquid
  • Color is colorless to light yellow
  • Ships at room temperature in continental US; may vary elsewhere
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-36248 Supplier Medchemexpress LLC Supplier No. HY126437K50ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly-L-lysine solution (0.01%, sterile-filtered, MW 70000-150000) is a biochemical reagent intended for research use only, and not for sale to patients. This solution is also referred to as PLL solution 0.01%, sterile-filtered, hydrobromide, MW 70000-150000, and is suitable for cell culture applications.

  • Molecular weight ranges from 70000 to 150000
  • Chemical formula is (C6H14N2O2)n.nBrH
  • CAS number is 25988-63-0
  • Appears as a liquid
  • Color is colorless to light yellow
  • Ships at room temperature in continental US; may vary elsewhere
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.