Learn More
Medchemexpress LLC Poly-L-lysine hydrobromide | 25988-63-0 | MFCD00165424 | 15,000-30,000 Da | C8H19BrN2O | 5 MG
SDP
Catalog No. 5000264405
Encompass_Preferred
Poly-L-lysine hydrobromide (MW 15,000-30,000 Da) is a cationic synthetic polyamino acid used to enhance cell adhesion and to form electrostatic complexes with negatively charged biomolecules. It is supplied as a water-soluble hydrobromide salt suitable for surface coating, biomaterials research, and gene delivery studies.
  • Cationic polymer that promotes cell attachment to substrates.
  • Water-soluble hydrobromide salt for easy aqueous preparation.
  • Molecular weight range 15,000-30,000 Da for moderate chain length properties.
  • Useful for cell culture coating, surface functionalization, and biomaterials research.
  • Forms electrostatic complexes with nucleic acids and proteins for delivery studies.
  • Available in small laboratory quantities for research applications.
Catalog No. 50-002-64405 Supplier Medchemexpress LLC Supplier No. HY1264375MG
Poly-L-lysine hydrobromide (MW 15,000-30,000 Da) is a cationic synthetic polyamino acid used to enhance cell adhesion and to form electrostatic complexes with negatively charged biomolecules. It is supplied as a water-soluble hydrobromide salt suitable for surface coating, biomaterials research, and gene delivery studies.
  • Cationic polymer that promotes cell attachment to substrates.
  • Water-soluble hydrobromide salt for easy aqueous preparation.
  • Molecular weight range 15,000-30,000 Da for moderate chain length properties.
  • Useful for cell culture coating, surface functionalization, and biomaterials research.
  • Forms electrostatic complexes with nucleic acids and proteins for delivery studies.
  • Available in small laboratory quantities for research applications.