Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-L-Lysine Hydrobromide, MP Biomedicals™

Catalog No. mp210269191 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210269191 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210269191 Supplier MP Biomedicals, Inc Supplier No. 0210269191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Promotes cell adhesion to solid substrates. Poly-L-Lysine Hydrobromide, MP Biomedicals is a positively charged amino acid polymer. It acts as a nonspecific attachment factor by enhancing electrostatic interactions between negatively charged ions of the ce

Poly-lysine binds to DNA, red cell membrane and any negatively charged protein. It is typically used as a coating substrate for culture dishes, slides, etc. It enhances electrostatic interactions to promote cell adhesion. With cells that can digest poly-L-lysine, poly-D-lysine should be used as the attachment factor.

  • When adsorbed to the culture surface, poly-lysine increases the number of positively charged sites available for cell binding
  • When it is absorbed to the cell culture surface, poly-L-lysine functions to increase the number of positively charged sites available for cell binding

Other applications of Poly-L-lysine include:

  • Conjugation to methotrexate for increased drug transport
  • Microencapsulation of islets
  • Use in a simple reproducible procedure for chromosomal preparations from a variety of tissues
  • Immobilation of living cells in biocompatible semipermeable microcapsules
  • In the preparation of polycationic beads
  • Conjugation to albumin and horseradish peroxidase to enhance cellular uptake
TRUSTED_SUSTAINABILITY

Specifications

CAS 25988-63-0
Content And Storage Store at –20°C
Format White Powder
Molecular Weight (g/mol) 402.54
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.