Learn More
Medchemexpress LLC Poly-L-lysine hydrochloride | 26124-78-7 | 30000-50000 | 100 MG
SDP
Catalog No. 5000435828
Encompass_Preferred
Poly-L-lysine hydrochloride is a nonspecific attachment factor for cells, useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. It also acts as a strong-attraction regulator, promoting liquid-liquid phase separation (LLPS) at low concentrations and suppressing it at high concentrations. It is also an antibacterial cationic peptide.
  • Nonspecific attachment factor for cells
  • Promotes cell adhesion to solid substrates
  • Enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface
  • Strong-attraction regulator
  • Promotes liquid-liquid phase separation (LLPS) at low concentrations
  • Suppresses LLPS at high concentrations
  • Antibacterial cationic peptide
Catalog No. 50-004-35828 Supplier Medchemexpress LLC Supplier No. HY126437A100MG
Poly-L-lysine hydrochloride is a nonspecific attachment factor for cells, useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. It also acts as a strong-attraction regulator, promoting liquid-liquid phase separation (LLPS) at low concentrations and suppressing it at high concentrations. It is also an antibacterial cationic peptide.
  • Nonspecific attachment factor for cells
  • Promotes cell adhesion to solid substrates
  • Enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface
  • Strong-attraction regulator
  • Promotes liquid-liquid phase separation (LLPS) at low concentrations
  • Suppresses LLPS at high concentrations
  • Antibacterial cationic peptide