Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Poly-L-Lysine Hydrochloride

Catalog No. 0215269090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-69090 500 mg
ICN15269080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-152-69090 Supplier MP Biomedicals™ Supplier No. 0215269090
Only null left
Add to Cart
Add to Cart

Poly-L-Lysine Hydrochloride is a cellular attachment factor in culture applications.

  • Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates.
  • Poly-L-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.
  • When adsorbed to the culture surface, poly-L-lysine increases the number of positively charged sites available for cell binding.
  • Polymers of both D- and L-lysine are used to coat solid surfaces.
  • Poly-L-lysine has been reported to improve the protein coating of ELISA plates.
TRUSTED_SUSTAINABILITY

Specifications

CAS 26124-78-7
Color Whitish
Content And Storage -20°C
Format Powder
For Use With (Application) Cellular attachment factor in culture applications
Molecular Weight (g/mol) >20,000 g/mol
Quantity 500 mg

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.