Learn More
Bioworld POLY-L- LYSINE SEPAROPORE
Catalog No. 50197862
Encompass
POLY-L- LYSINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: Poly-L-Lysine.; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow sp...
Catalog No. 50-197-862 Supplier Bioworld Supplier No. 201810752
POLY-L- LYSINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: Poly-L-Lysine.; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow sp...