Learn More
Ted Pella Inc POLY L LYSINE SOL 10 ML
SDP
Catalog No. NC9803810
Encompass
Poly-L-Lysine solution 10mL Surface Coating for adhesion of cells/particles 0.1% w/v aqueous solution preservative for tissue sections on glass slides
Catalog No. NC9803810 Supplier Ted Pella Inc Supplier No. 18026
Poly-L-Lysine solution 10mL Surface Coating for adhesion of cells/particles 0.1% w/v aqueous solution preservative for tissue sections on glass slides