Learn More
TRIBIOSCIENCE INC Poly-L-Lysine Solution (0.01%) in PBS, Ready to Use, 50ML
SDP
Catalog No. NC3834048
Encompass
Poly-L-Lysine is widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces. This molecule has been used to culture a wide variety of cell types. This product’s molecular weight ranges from 70,000 to 150,000 Da yielding a solution viscosity for easy handling while providing sufficient binding sites for cell attachment.
Catalog No. NC3834048 Supplier TRIBIOSCIENCE INC Supplier No. TBS804050ML
Poly-L-Lysine is widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces. This molecule has been used to culture a wide variety of cell types. This product’s molecular weight ranges from 70,000 to 150,000 Da yielding a solution viscosity for easy handling while providing sufficient binding sites for cell attachment.