Learn More
Supply Solutions Poly-L-lysine solution, 25988-63-0, MFCD00165424, 50mL
Supplier:  Supply Solutions SIGMA P483250ML
Encompass_Preferred
mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, Synonym: PLL Solution; Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.
Catalog No. 50-161-3347