Learn More
Sigma Aldrich Fine Chemicals Biosciences POLY-L-LYSINE SOLUTION, 50ML
Catalog No. NC0818114
Encompass
Components Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.
Catalog No. NC0818114 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P470750ML
Components Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.